1-(N-Boc)-3-cyanoazetidine
Supplier: Apollo Scientific
Warning
OR2250-25G OR2250-5G
APOSOR2250-25GEA 133 EUR
APOSOR2250-25G APOSOR2250-5G
1-(N-Boc)-3-cyanoazetidine
1-(N-Boc)-3-cyanoazetidine
Formula: C₉H₁₄N₂O₂
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD06796640
CAS Number: 142253-54-1