6-Methylindole
Supplier: Apollo Scientific
Warning
Synonyms: 6-Methyl-1H-indole
OR2254-5G OR2254-25G
APOSOR2254-5GEA 26.1 EUR
APOSOR2254-5G APOSOR2254-25G
6-Methylindole
6-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 112 °C/5mmHg
Melting Pt: 29…32 °C
Density: 1.059 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005682
CAS Number: 3420-02-8
UN: 3335
ADR: 9,III