4-Decylaniline hydrochloride
Supplier: Apollo Scientific
Warning
OR22593-10G OR22593-1G OR22593-5G
APOSOR22593-10GEA 346 EUR
APOSOR22593-10G APOSOR22593-1G APOSOR22593-5G
4-Decylaniline hydrochloride
4-Decylaniline hydrochloride
Formula: C₁₆H₂₈ClN
Storage Temperature: Refrigerator
MDL Number: MFCD00155303
CAS Number: 175136-55-7