4-Bromo-2-methoxyphenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromoguaiacol, 5-Brom-2-hydroxyanisole
OR22659-25G OR22659-5G OR22659-1G
APOSOR22659-25GEA 87 EUR
APOSOR22659-25G APOSOR22659-5G APOSOR22659-1G
4-Bromo-2-methoxyphenol 97%
4-Bromo-2-methoxyphenol
Formula: C₇H₇BrO₂
MW: 203.04 g/mol
Boiling Pt: 129…132 °C/12mmHg
Melting Pt: 34…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00051937
CAS Number: 7368-78-7