3-(Benzyloxy)-4,5-dimethoxybenzaldehyde
Supplier: Apollo Scientific
Warning
OR22667-1G OR22667-250MG
APOSOR22667-1GEA 398 EUR
APOSOR22667-1G APOSOR22667-250MG
3-(Benzyloxy)-4,5-dimethoxybenzaldehyde
3-(Benzyloxy)-4,5-dimethoxybenzaldehyde
Formula: C₁₆H₁₆O₄
Melting Pt: 138…141 °C
Storage Temperature: Ambient
CAS Number: 57230-04-3