4,6-Dichloropyrimidine 98%
Supplier: Apollo Scientific
|
Danger
|
OR22727-100G
OR22727-25G
OR22727-500G
APOSOR22727-100GEA
43.5
EUR
APOSOR22727-100G
APOSOR22727-25G
APOSOR22727-500G
4,6-Dichloropyrimidine 98%
4,6-Dichloropyrimidine
Formula:
C₄H₂Cl₂N₂ MW: 148.98 g/mol Boiling Pt: 176 °C Melting Pt: 64…68 °C Density: 1.734 Storage Temperature: Ambient |
MDL Number:
MFCD00006109 CAS Number: 1193-21-1 EINECS: 214-770-2 UN: 3263 ADR: 8 II |