6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
Supplier: Apollo Scientific
Warning
OR22802-1G OR22802-5G OR22802-25G
APOSOR22802-1GEA 21.9 EUR
APOSOR22802-1G APOSOR22802-5G APOSOR22802-25G
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione
Formula: C₃H₂BrN₃O₂
MW: 191.98 g/mol
Melting Pt: 232…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00154757
CAS Number: 4956-05-2