2,4,6-Trichloropyridine 98%
Supplier: Apollo Scientific
Danger
OR22840-25G OR22840-5G OR22840-100G
APOSOR22840-25GEA 132 EUR
APOSOR22840-25G APOSOR22840-5G APOSOR22840-100G
2,4,6-Trichloropyridine 98%
2,4,6-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Boiling Pt: 88…90 °C/10mmHg
Melting Pt: 32…37 °C
Density: 1.595 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00971928
CAS Number: 16063-69-7