Tropine
Supplier: Apollo Scientific
Warning
Synonyms: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
OR22908-100G OR22908-500G
APOSOR22908-100GEA 72.5 EUR
APOSOR22908-100G APOSOR22908-500G
Tropine
Tropine
Formula: C₈H₁₅NO
MW: 141.21 g/mol
Boiling Pt: 232…234 °C
Melting Pt: 61…65 °C
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2