(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one
OR22930-100G OR22930-1KG OR22930-500G
APOSOR22930-100GEA 112 EUR
APOSOR22930-100G APOSOR22930-1KG APOSOR22930-500G
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97+%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
MW: 109.1259 g/mol
Boiling Pt: 102…106 °C/0.25mmHg
Melting Pt: 54…58 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00213364
CAS Number: 49805-30-3
EINECS: 421-830-0