6-Bromoacetyl-1,4-benzodioxane
Supplier: Apollo Scientific
Danger
Synonyms: 6-(Bromoacetyl)-2,3-dihydro-1,4-benzodioxine
OR23006-1G OR23006-250MG OR23006-5G
APOSOR23006-1GEA 109 EUR
APOSOR23006-1G APOSOR23006-250MG APOSOR23006-5G
6-Bromoacetyl-1,4-benzodioxane
6-Bromoacetyl-1,4-benzodioxane
Formula: C₁₀H₉BrO₃
Melting Pt: 114…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00099402
CAS Number: 4629-54-3
UN: 1759
ADR: 8,III