2-Furoyl chloride
Supplier: Apollo Scientific
Danger
OR23022-250G OR23022-50G
APOSOR23022-250GEA 201 EUR
APOSOR23022-250G APOSOR23022-50G
2-Furoyl chloride
2-Furoyl chloride
Formula: C₅H₃ClO₂
MW: 130.53 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –2 °C
Density: 1.325 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00003228
CAS Number: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8,II