2-Furaldehyde
Supplier: Apollo Scientific
|
Danger
|
OR23023-1KG
OR23023-250G
APOSOR23023-1KGEA
101
EUR
APOSOR23023-1KG
APOSOR23023-250G
2-Furaldehyde
2-Furaldehyde
Formula:
C₅H₄O₂ Boiling Pt: 159…162 °C Melting Pt: –38 °C Density: 1.159 g/cm³ (25 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003229 CAS Number: 98-01-1 EINECS: 202-627-7 UN: 1199 ADR: 6.1+3,II |