Furfuryl alcohol
Supplier: Apollo Scientific
Synonyms: Furfurol
OR23024-1KG OR23024-250G
APOSOR23024-1KGEA 179 EUR
APOSOR23024-1KG APOSOR23024-250G
Furfuryl alcohol
Furfuryl alcohol
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 170 °C
Melting Pt: –29 °C
Density: 1.135 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
UN: 2874
ADR: 6.1 III
Merck Index: 13,04326