3-Furaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: Furan-3-carbaldehyde
OR23028-1G OR23028-10G
APOSOR23028-1GEA 29 EUR
APOSOR23028-1G APOSOR23028-10G
3-Furaldehyde 98%
3-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 145 °C
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010424
CAS Number: 498-60-2
UN: 1199
ADR: 6.1+3 II