3-Furanmethanol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Furyl Alcohol, 3-(Hydroxymethyl)furan
OR23029-25G
APOSOR23029-25GEA 213 EUR
APOSOR23029-25G
3-Furanmethanol
3-Furanmethanol
Formula: C₅H₆O₂
MW: 98.10 g/mol
Boiling Pt: 79…80 °C/17mmHg
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005352
CAS Number: 4412-91-3
UN: 1987
ADR: 3,III