3-Bromofuran
Supplier: Apollo Scientific
Danger
OR23030-100G OR23030-25G OR23030-5G
APOSOR23030-100GEA 400 EUR
APOSOR23030-100G APOSOR23030-25G APOSOR23030-5G
3-Bromofuran
3-Bromofuran
Formula: C₄H₃BrO
MW: 146.97 g/mol
Boiling Pt: 102…104 °C
Density: 1.635 g/cm³ (25 °C)
Flash Pt: 3 °C
Storage Temperature: Freezer
MDL Number: MFCD00005347
CAS Number: 22037-28-1
EINECS: 244-744-6
UN: 1993
ADR: 3 II