1-Methylpyrrole-2-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-1H-pyrrole-2-carboxylic acid, 1-Methyl-2-pyrrolecarboxylic acid
OR23038-1G OR23038-5G
APOSOR23038-1GEA 24.8 EUR
APOSOR23038-1G APOSOR23038-5G
1-Methylpyrrole-2-carboxylic acid
1-Methylpyrrole-2-carboxylic acid
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 131…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003088
CAS Number: 6973-60-0
EINECS: 230-217-8