3-Bromopyridine 99%
Supplier: Apollo Scientific
Danger
OR23062-100G OR23062-25G OR23062-500G
APOSOR23062-100GEA 29 EUR
APOSOR23062-100G APOSOR23062-25G APOSOR23062-500G
3-Bromopyridine 99%
3-Bromopyridine
Formula: C₅H₄BrN
Boiling Pt: 172…174 °C
Melting Pt: –27 °C
Density: 1.64 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2929
ADR: 6.1+3,II