4-Bromopyridinium chloride 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride
OR23067-100G OR23067-25G OR23067-500G
APOSOR23067-100GEA 69.6 EUR
APOSOR23067-100G APOSOR23067-25G APOSOR23067-500G
4-Bromopyridinium chloride 95+%
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
MW: 194.46 g/mol
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4