4-(Aminomethyl)pyridine
Supplier: Apollo Scientific
Danger
Synonyms: 4-Picolamine, (4-Pyridyl)methylamine, Pyridin-4-ylmethanamine, 4-Picolylamine, 4-Pyridylmethylamine
OR23068-100G OR23068-25G OR23068-5G
APOSOR23068-100GEA 97.3 EUR
APOSOR23068-100G APOSOR23068-25G APOSOR23068-5G
4-(Aminomethyl)pyridine
4-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 228…230 °C
Melting Pt: –8 °C
Density: 1.065 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006449
CAS Number: 3731-53-1
EINECS: 223-092-6
UN: 2735
ADR: 8 II