6-Quinolinecarboxylic acid 98%
Supplier: Apollo Scientific
Warning
OR23075-10G OR23075-1G OR23075-25G
APOSOR23075-10GEA 50.9 EUR
APOSOR23075-10G APOSOR23075-1G APOSOR23075-25G
6-Quinolinecarboxylic acid 98%
6-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345
Melting Pt: 293…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00047613
CAS Number: 10349-57-2
EINECS: 233-761-4