4-Quinolinecarbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde
OR23077-25G OR23077-5G OR23077-1G
APOSOR23077-25GEA 184 EUR
APOSOR23077-25G APOSOR23077-5G APOSOR23077-1G
4-Quinolinecarbaldehyde
4-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 45…52 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0