1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Bromoacetyl)-1,3-benzothiazole
OR23089-1G OR23089-250MG
APOSOR23089-1GEA 139 EUR
APOSOR23089-1G APOSOR23089-250MG
1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone
1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone
Formula: C₉H₆BrNOS
MW: 256.13 g/mol
Melting Pt: 86…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03659698
CAS Number: 54223-20-0
UN: 1759
ADR: 8 III