2-Pyrrolecarbaldehyde 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde
OR23112-100G OR23112-25G OR23112-500G
APOSOR23112-100GEA 102 EUR
APOSOR23112-100G APOSOR23112-25G APOSOR23112-500G
2-Pyrrolecarbaldehyde 99%
2-Pyrrolecarbaldehyde
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 217…219 °C
Melting Pt: 43…46 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005217
CAS Number: 1003-29-8
EINECS: 213-705-5