(1H-Imidazol-2-yl)methanol hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Hydroxymethyl)-1H-imidazole hydrochloride
OR23115-1G OR23115-5G
APOSOR23115-1GEA 319 EUR
APOSOR23115-1G APOSOR23115-5G
(1H-Imidazol-2-yl)methanol hydrochloride 97%
(1H-Imidazol-2-yl)methanol hydrochloride
Formula: C₄H₇ClN₂O
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD03659700
CAS Number: 116177-22-1