3-Bromomethyl-5-methyl-isoxazole
Supplier: Apollo Scientific
Danger
OR23143-1G OR23143-250MG OR23143-5G
APOSOR23143-1GEA 87 EUR
APOSOR23143-1G APOSOR23143-250MG APOSOR23143-5G
3-Bromomethyl-5-methyl-isoxazole
3-Bromomethyl-5-methyl-isoxazole
Formula: C₅H₆BrNO
MW: 176.02 g/mol
Boiling Pt: 44 °C/0.4mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD02677720
CAS Number: 130628-75-0
UN: 3265
ADR: 8,III