1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride
Supplier: Apollo Scientific
Danger
OR23189-250MG OR23189-1G OR23189-5G
APOSOR23189-250MGEA 43.5 EUR
APOSOR23189-250MG APOSOR23189-1G APOSOR23189-5G
1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride
1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride
Formula: C₆H₇ClN₂O
Melting Pt: 73…74 °C
Storage Temperature: Ambient
MDL Number: MFCD03407950
CAS Number: 49783-84-8
UN: 3261
ADR: 8,III