(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,5-Dimethyl-3-(hydroxymethyl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazol-3-yl-methanol
OR23190-1G OR23190-5G
APOSOR23190-1GEA 71.2 EUR
APOSOR23190-1G APOSOR23190-5G
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol
Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
Boiling Pt: 118 °C/2mmHg
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD03659702
CAS Number: 153912-60-8