2-(2-Bromoacetyl)thiophene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-(thiophen-2-yl)ethanone, 2-(Bromoacetyl)thiophene
OR23196-10G OR23196-1G OR23196-5G
APOSOR23196-10GEA 154 EUR
APOSOR23196-10G APOSOR23196-1G APOSOR23196-5G
2-(2-Bromoacetyl)thiophene 97%
2-(2-Bromoacetyl)thiophene
Formula: C₆H₅BrOS
MW: 205.07 g/mol
Melting Pt: 29…33 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD02677721
CAS Number: 10531-41-6
EINECS: 206-994-4
UN: 3261
ADR: 8 II