(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Aminomethyl)-1,3,5-trimethyl-1H-pyrazole
OR23240-250MG OR23240-1G
APOSOR23240-250MGEA 144 EUR
APOSOR23240-250MG APOSOR23240-1G
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine 97%
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine
Formula: C₇H₁₃N₃
MW: 139.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02055811
CAS Number: 352018-93-0
UN: 2735
ADR: 8,III