4-(1,2,3-Thiadiazol-4-yl)benzoic acid
Supplier: Apollo Scientific
Warning
OR23259-1G OR23259-250MG
APOSOR23259-1GEA 236 EUR
APOSOR23259-1G APOSOR23259-250MG
4-(1,2,3-Thiadiazol-4-yl)benzoic acid
4-(1,2,3-Thiadiazol-4-yl)benzoic acid
Formula: C₉H₆N₂O₂S
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00085167
CAS Number: 187999-31-1