(4-Pyrazol-1-ylphenyl)methanol
Supplier: Apollo Scientific
Warning
Synonyms: (4-Pyrazol-1-yl-phenyl)methanol
OR23294-1G OR23294-25G OR23294-5G
APOSOR23294-1GEA 21.9 EUR
APOSOR23294-1G APOSOR23294-25G APOSOR23294-5G
(4-Pyrazol-1-ylphenyl)methanol
(4-Pyrazol-1-ylphenyl)methanol
Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
Melting Pt: 76…87 °C
Storage Temperature: Ambient
MDL Number: MFCD02682057
CAS Number: 143426-49-7