1-(4-Iodophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR23295-1G OR23295-5G
APOSOR23295-1GEA 116 EUR
APOSOR23295-1G APOSOR23295-5G
1-(4-Iodophenyl)-1H-pyrazole
1-(4-Iodophenyl)-1H-pyrazole
Formula: C₉H₇IN₂
MW: 270.0700 g/mol
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD02681941
CAS Number: 368869-86-7