(4-(1H-Pyrazol-1-yl)phenyl)methanamine
Supplier: Apollo Scientific
Danger
OR23296-1G OR23296-250MG OR23296-5G
APOSOR23296-1GEA 137 EUR
APOSOR23296-1G APOSOR23296-250MG APOSOR23296-5G
(4-(1H-Pyrazol-1-yl)phenyl)methanamine
(4-(1H-Pyrazol-1-yl)phenyl)methanamine
Formula: C₁₀H₁₁N₃
MW: 173.2144 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD02682058
CAS Number: 368870-03-5
UN: 3259
ADR: 8 III