4-(1H-Pyrazol-1-yl)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 4-Pyrazol-1-yl-phenylamine
OR23297-1G OR23297-25G OR23297-5G
APOSOR23297-1GEA 21.9 EUR
APOSOR23297-1G APOSOR23297-25G APOSOR23297-5G
4-(1H-Pyrazol-1-yl)aniline
4-(1H-Pyrazol-1-yl)aniline
Formula: C₉H₉N₃
MW: 159.1879 g/mol
Melting Pt: 54…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00626298
CAS Number: 17635-45-9