2-Methyl-4-phenylpyrimidine-5-carboxylic acid
Supplier: Apollo Scientific
Warning
OR23330-1G OR23330-250MG
APOSOR23330-1GEA 98.6 EUR
APOSOR23330-1G APOSOR23330-250MG
2-Methyl-4-phenylpyrimidine-5-carboxylic acid
2-Methyl-4-phenylpyrimidine-5-carboxylic acid
Formula: C₁₂H₁₀N₂O₂
MW: 214.23 g/mol
Melting Pt: 210…211 °C
Storage Temperature: Ambient
MDL Number: MFCD02682072
CAS Number: 127958-10-5