tert-Butyl nitrite 95%
Supplier: Apollo Scientific
Danger
OR2337-100G OR2337-25G OR2337-500G
APOSOR2337-100GEA 71.2 EUR
APOSOR2337-100G APOSOR2337-25G APOSOR2337-500G
tert-Butyl nitrite 95%
tert-Butyl nitrite
Formula: C₄H₉NO₂
Boiling Pt: 61…63 °C
Density: 0.867 g/cm³ (25 °C)
Flash Pt: –10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002055
CAS Number: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3,II