ω-Bromo-4-methoxyacetophenone
Supplier: Apollo Scientific
Danger
OR23418-100G OR23418-25G OR23418-5G
APOSOR23418-100GEA 73 EUR
APOSOR23418-100G APOSOR23418-25G APOSOR23418-5G
ω-Bromo-4-methoxyacetophenone
ω-Bromo-4-methoxyacetophenone
Formula: C₉H₉BrO₂
Melting Pt: 69…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000201
CAS Number: 2632-13-5
EINECS: 220-118-8
UN: 3261
ADR: 8,II