1,10-Phenanthroline monohydrate
Supplier: Apollo Scientific
Danger
Synonyms: 1,10-Phenanthroline hydrate
OR2359-100G OR2359-250G OR2359-25G
APOSOR2359-100GEA 58 EUR
APOSOR2359-100G APOSOR2359-250G APOSOR2359-25G
1,10-Phenanthroline monohydrate
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 97…104 °C
Density: 1.259 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III