Methyl-2-iodobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-iodobenzoate, 2-Iodobenzoic acid methyl ester
OR23611-500G
APOSOR23611-500GEA 89.9 EUR
APOSOR23611-500G
Methyl-2-iodobenzoate 98%
Methyl 2-iodobenzoate
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 148…150 °C/10mmHg
Density: 1.784 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016351
CAS Number: 610-97-9
EINECS: 210-243-6