Methyl-2-bromo-5-methoxybenzoate
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 2-bromo-5-methoxybenzoate, 2-Bromo-5-methoxybenzoic acid methyl ester
OR23647-100G OR23647-25G OR23647-10G
APOSOR23647-100GEA 415 EUR
APOSOR23647-100G APOSOR23647-25G APOSOR23647-10G
Methyl-2-bromo-5-methoxybenzoate
Methyl 2-bromo-5-methoxybenzoate
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 271 °C
Density: 1.52 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051594
CAS Number: 35450-36-3
UN: 2810
ADR: 6.1,III