4-Methyl-1,2,3-thiadiazole-5-carbohydrazide
Supplier: Apollo Scientific
Warning
OR23650-1G OR23650-5G
APOSOR23650-1GEA 111 EUR
APOSOR23650-1G APOSOR23650-5G
4-Methyl-1,2,3-thiadiazole-5-carbohydrazide
4-Methyl-1,2,3-thiadiazole-5-carbohydrazide
Formula: C₄H₆N₄OS
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00052212
CAS Number: 75423-15-3