4,6-Dibromo-2,3-dichloroaniline
Supplier: Apollo Scientific
Warning
OR23757-100G OR23757-25G OR23757-500G
APOSOR23757-100GEA 244 EUR
APOSOR23757-100G APOSOR23757-25G APOSOR23757-500G
4,6-Dibromo-2,3-dichloroaniline
4,6-Dibromo-2,3-dichloroaniline
Formula: C₆H₃Br₂Cl₂N
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00052815
CAS Number: 113571-15-6