2,3-Dibromothiophene 98%
Supplier: Apollo Scientific
Warning
OR23788-25G OR23788-5G
APOSOR23788-25GEA 24.8 EUR
APOSOR23788-25G APOSOR23788-5G
2,3-Dibromothiophene 98%
2,3-Dibromothiophene
Formula: C₄H₂Br₂S
Boiling Pt: 214…216 °C
Density: 2.137 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00005418
CAS Number: 3140-93-0
EINECS: 221-542-6
UN: 1993
ADR: 3,III