α,3,5-Tribromotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
OR23789-25G
APOSOR23789-25GEA 59.6 EUR
APOSOR23789-25G
α,3,5-Tribromotoluene 98%
α,3,5-Tribromotoluene
Formula: C₇H₅Br₃
MW: 328.83 g/mol
Boiling Pt: 169 °C/15mmHg
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00052415
CAS Number: 56908-88-4
UN: 3261
ADR: 8,II