2,4,6-Trimethylheptane
Supplier: Apollo Scientific
Warning
OR2379-1G OR2379-250MG
APOSOR2379-1GEA 480 EUR
APOSOR2379-1G APOSOR2379-250MG
2,4,6-Trimethylheptane
2,4,6-Trimethylheptane
Formula: C₁₀H₂₂
Boiling Pt: 144…147 °C
Density: 0.722 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 2613-61-8
UN: 3295
ADR: 3 III