3,5-Di-tert-butylaniline
Supplier: Apollo Scientific
Warning
OR2387-1G OR2387-5G
APOSOR2387-1GEA 71.2 EUR
APOSOR2387-1G APOSOR2387-5G
3,5-Di-tert-butylaniline
3,5-Di-tert-butylaniline
Formula: C₁₄H₂₃N
Melting Pt: 54…57 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134096
CAS Number: 2380-36-1