3-(Dimethylamino)-2-(2-nitrophenyl)acrylaldehyde
Supplier: Apollo Scientific
OR23877-1G OR23877-5G OR23877-10G
APOSOR23877-1GEA 111 EUR
APOSOR23877-1G APOSOR23877-5G APOSOR23877-10G
3-(Dimethylamino)-2-(2-nitrophenyl)acrylaldehyde
3-(Dimethylamino)-2-(2-nitrophenyl)acrylaldehyde
Formula: C₁₁H₁₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22566373
CAS Number: 53868-36-3