1,4-Dimethoxynaphthalene 96%
Supplier: Apollo Scientific
Warning
OR23941-1G OR23941-25G OR23941-5G
APOSOR23941-1GEA 60.1 EUR
APOSOR23941-1G APOSOR23941-25G APOSOR23941-5G
1,4-Dimethoxynaphthalene 96%
1,4-Dimethoxynaphthalene
Formula: C₁₂H₁₂O₂
Melting Pt: 84…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052378
CAS Number: 10075-62-4
EINECS: 233-209-2